Anti-CD10 Mouse Monoclonal Antibody [Clone: 56C6]

Supplier: ANTIBODIES.COM LLC
A254284-100 A254284-500
76968-558EA 603.88 CAD
76968-558 76953-584
Anti-CD10 Mouse Monoclonal Antibody [Clone: 56C6]
Antibodies
Mouse monoclonal [56C6] antibody to CD10 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1:50 to 1:100

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: 56C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR