Anti-B MyB Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A96087-100
77052-896EA 765.04 CAD
77052-896
Anti-B MyB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to B-Myb for WB, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IF, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:20000

Type: Primary
Antigen: B MyB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR