Anti-beta Actin Rabbit Monoclonal Antibody [Clone: ARC5115-01] (HRP (Horseradish Peroxidase))

Supplier: ANTIBODIES.COM LLC
A92928-100
77041-982EA 355.5 CAD
77041-982 77041-980
Anti-beta Actin Rabbit Monoclonal Antibody [Clone: ARC5115-01] (HRP (Horseradish Peroxidase))
Antibodies
Rabbit monoclonal [ARC5115-01] antibody to beta Actin for WB with samples derived from Human, Mouse, Rat, Chicken, Zebrafish and Porcine.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:2, 000-1:50, 000

Type: Primary
Antigen: beta Actin
Clonality: Monoclonal
Clone: ARC5115-01
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Zebrafish, Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR