EZBio® Titanium Round (TR) Polypropylene Carboys
Supplier: Foxx Life Sciences
515-CF51-FLS 515-CF52-FLS 515-CG51-FLS 515-CG52-FLS 515-CH51-FLS 515-CH52-FLS
77296-506EA 188.69 CAD
77296-506 77296-512 77296-508 77296-514 77296-510 77296-516
EZBio® Titanium Round (TR) Polypropylene Carboys
Bottles Carboys
EZBio® TR carboys are available in sizes 5, 10 and 20 L. They are available in non sterile and sterile formats.
  • Available in HDPE, PC, PETG, and PP; FDA 21 CFR 177 grade materials
  • Sanitary tops are available
  • Molded with ultra-pure USP Class VI resins
  • EZBio® TR carboys are cleaned, triple inspected, and double bagged
  • Patented 83B VersaCap®
  • Manufactured in an ISO 13485 facility in North America
Foxx Life Sciences' EZBio® Titanium Round (TR) carboys are advanced fluid handling systems developed with the end user in mind. The EZBio® TR carboy serves as a highly versatile container suitable for all fluid management applications. The EZBio® TR carboys are molded with ultra-pure USP Class VI resins, FDA 21 CFR 177 grade materials, and are available double bagged, sterile or non sterile.
Certifications: UPS Class VI materials.
Packaging: Double bagged.