Anti-Thomsen-Friedenreich Antigen Mouse Monoclonal Antibody [clone: A78-G/A7]

Supplier: Antibodies.com LLC
A251089-100
76961-856EA 854.39 CAD
76961-856
Anti-Thomsen-Friedenreich Antigen Mouse Monoclonal Antibody [clone: A78-G/A7]
Antibodies
Mouse monoclonal [A78-G/A7] antibody to Thomsen-Friedenreich Antigen for IF and IHC-P with samples derived from Human, Mouse and Rat.

  • Validated Applications: IF, IHC-P
  • Recommended Dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Thomsen-Friedenreich Antigen
Clonality: Monoclonal
Clone: A78-G/A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR