Anti-alpha 2 Macroglobulin Mouse Monoclonal Antibody [clone: A2M/3621]

Supplier: Antibodies.com LLC
A248796-100
76955-298EA 854.39 CAD
76955-298
Anti-alpha 2 Macroglobulin Mouse Monoclonal Antibody [clone: A2M/3621]
Antibodies
Mouse monoclonal [A2M/3621] antibody to alpha 2 Macroglobulin for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: alpha 2 Macroglobulin
Clonality: Monoclonal
Clone: A2M/3621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR