Anti-GDF8 Mouse Monoclonal Antibody [clone: GDF9/4261]

Supplier: ANTIBODIES.COM LLC
A248702-100
76960-054EA 877.69 CAD
76960-054
Anti-GDF8 Mouse Monoclonal Antibody [clone: GDF9/4261]
Antibodies
Mouse monoclonal [GDF9/4261] antibody to GDF8 for ELISA and WB with samples derived from Human.

  • Validated Applications: ELISA, WB
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: GDF8
Clonality: Monoclonal
Clone: GDF9/4261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR