Anti-CPS1 Mouse Monoclonal Antibody [clone: CPS1/1022]

Supplier: ANTIBODIES.COM LLC
A248255-100
76958-272EA 877.69 CAD
76958-272
Anti-CPS1 Mouse Monoclonal Antibody [clone: CPS1/1022]
Antibodies
Mouse monoclonal [CPS1/1022] antibody to CPS1 for IHC-P with samples derived from Human and Canine.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CPS1
Clonality: Monoclonal
Clone: CPS1/1022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR