Anti-V5 Tag Goat Polyclonal Antibody

Supplier: Antibodies.com
A121614-15 A121614-600
76835-106EA 1191.79 CAD
76835-106 76834-334
Anti-V5 Tag Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to V5 Tag for WB.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:5,000, IF: 1:500 to 1:2,000
  • Host: Goat
  • Clonality: Polyclonal
  • Isotype: IgG
  • Conjugate: Unconjugated

Type: Primary
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR