Anti-Tyrosine Hydroxylase Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A104319-50 A104319-100
77074-216EA 676.4 CAD
77074-216 77077-696
Anti-Tyrosine Hydroxylase Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Tyrosine Hydroxylase for WB and ICC/IF with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF
  • Recommended Dilutions: WB: 1:5,000, ICC/IF: 1:10,000

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR