Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: 4H2]

Supplier: ANTIBODIES.COM LLC
A104315-50 A104315-100
77066-416EA 676.4 CAD
77066-416 77071-344
Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: 4H2]
Antibodies
Mouse monoclonal (4H2) antibody to Tyrosine Hydroxylase for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.

  • Validated Applications: WB, ICC/IF, IHC
  • Recommended Dilutions: WB: 1:1,000 to 1:2,000, ICC/IF: 1:1,000, IHC: 1:1,000

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR