Anti-CD3E Antigen Mouse Monoclonal Antibody [clone: ABT-CD3]

Supplier: ANTIBODIES.COM LLC
A100704-100 A100704-50
77066-496EA 765.04 CAD
77066-496 77066-394
Anti-CD3E Antigen Mouse Monoclonal Antibody [clone: ABT-CD3]
Antibodies
Mouse monoclonal (ABT-CD3) antibody to CD3 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD3E Antigen
Clonality: Monoclonal
Clone: ABT-CD3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR