Anti-SEC16A Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A100215-100
77078-562EA 765.04 CAD
77078-562
Anti-SEC16A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SEC16A for IHC and ELISA with samples derived from Human.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000

Type: Primary
Antigen: SEC16A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR