Anti-CD1a Mouse Monoclonal Antibody [clone: ABT-CD1a]

Supplier: ANTIBODIES.COM LLC
A100036-50 A100036-100
77072-620EA 563.74 CAD
77072-620 77065-764
Anti-CD1a Mouse Monoclonal Antibody [clone: ABT-CD1a]
Antibodies
Mouse monoclonal (ABT-CD1a) antibody to CD1a for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: ABT-CD1a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR