Anti-CD99R Mouse Monoclonal Antibody [clone: ABT-CD99]

Supplier: ANTIBODIES.COM LLC
A100022-50 A100022-100
77077-258EA 563.74 CAD
77077-258 77061-560
Anti-CD99R Mouse Monoclonal Antibody [clone: ABT-CD99]
Antibodies
Mouse monoclonal (ABT-CD99) antibody to CD99 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD99R
Clonality: Monoclonal
Clone: ABT-CD99
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR