Anti-Proinsulin Mouse Monoclonal Antibody [clone: ABT-INS]

Supplier: ANTIBODIES.COM LLC
A99499-50 A99499-100
77066-760EA 563.74 CAD
77066-760 77075-740
Anti-Proinsulin Mouse Monoclonal Antibody [clone: ABT-INS]
Antibodies
Mouse monoclonal (ABT-INS) antibody to Insulin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Proinsulin
Clonality: Monoclonal
Clone: ABT-INS
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR