Anti-p57 Kip2 Mouse Monoclonal Antibody [clone: ABT-P57]

Supplier: ANTIBODIES.COM LLC
A99497-100 A99497-50
77062-294EA 765.04 CAD
77062-294 77080-630
Anti-p57 Kip2 Mouse Monoclonal Antibody [clone: ABT-P57]
Antibodies
Mouse monoclonal (ABT-P57) antibody to p57kip2 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: p57 Kip2
Clonality: Monoclonal
Clone: ABT-P57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR