Anti-Gastrin I Mouse Monoclonal Antibody [clone: ABT-GAST]

Supplier: ANTIBODIES.COM LLC
A99260-100 A99260-50
77064-298EA 765.04 CAD
77064-298 77072-536
Anti-Gastrin I Mouse Monoclonal Antibody [clone: ABT-GAST]
Antibodies
Mouse monoclonal (ABT-GAST) antibody to Gastrin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Gastrin I
Clonality: Monoclonal
Clone: ABT-GAST
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR