Anti-CD44v6 Mouse Monoclonal Antibody [clone: ABT-CD44]

Supplier: ANTIBODIES.COM LLC
A99231-100 A99231-50
77067-706EA 765.04 CAD
77067-706 77064-362
Anti-CD44v6 Mouse Monoclonal Antibody [clone: ABT-CD44]
Antibodies
Mouse monoclonal (ABT-CD44) antibody to CD44 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: CD44v6
Clonality: Monoclonal
Clone: ABT-CD44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR