Anti-PRPF31 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A99007-100
77068-144EA 765.04 CAD
77068-144
Anti-PRPF31 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PRP31 for WB and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: PRPF31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR