Anti-UBTD2 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A98949-100
77079-978EA 765.04 CAD
77079-978
Anti-UBTD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to UBTD2 for WB and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR