Anti-hCG beta Mouse Monoclonal Antibody [clone: ABT-HCGβ]

Supplier: ANTIBODIES.COM LLC
A98913-100 A98913-50
77079-594EA 765.04 CAD
77079-594 77070-954
Anti-hCG beta Mouse Monoclonal Antibody [clone: ABT-HCGβ]
Antibodies
Mouse monoclonal (ABT-HCGβ) antibody to hCG beta for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: ABT-HCGβ
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR