Anti-Tyrosinase Mouse Monoclonal Antibody [clone: ABT-TYR]

Supplier: ANTIBODIES.COM LLC
A98700-100 A98700-50
77076-682EA 765.04 CAD
77076-682 77080-798
Anti-Tyrosinase Mouse Monoclonal Antibody [clone: ABT-TYR]
Antibodies
Mouse monoclonal (ABT-TYR) antibody to Tyrosinase for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Tyrosinase
Clonality: Monoclonal
Clone: ABT-TYR
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR