Anti-AMACR Mouse Monoclonal Antibody [clone: ABT-AMACR]

Supplier: ANTIBODIES.COM LLC
A98671-100 A98671-50
77061-574EA 765.04 CAD
77061-574 77067-428
Anti-AMACR Mouse Monoclonal Antibody [clone: ABT-AMACR]
Antibodies
Mouse monoclonal (ABT-AMACR) antibody to AMACR for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: AMACR
Clonality: Monoclonal
Clone: ABT-AMACR
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR