Anti-ZAP70 Mouse Monoclonal Antibody [clone: ABT-ZAP70]

Supplier: ANTIBODIES.COM LLC
A98669-100 A98669-50
77068-228EA 765.04 CAD
77068-228 77067-622
Anti-ZAP70 Mouse Monoclonal Antibody [clone: ABT-ZAP70]
Antibodies
Mouse monoclonal (ABT-ZAP70) antibody to ZAP70 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: ABT-ZAP70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR