Anti-Cytokeratin 4 + 5 + 6 + 8 + 10 + 13 + 18 Mouse Monoclonal Antibody [clone: ABT-CK4]

Supplier: ANTIBODIES.COM LLC
A97803-100 A97803-50
77059-766EA 765.04 CAD
77059-766 77059-768
Anti-Cytokeratin 4 + 5 + 6 + 8 + 10 + 13 + 18 Mouse Monoclonal Antibody [clone: ABT-CK4]
Antibodies
Mouse monoclonal (ABT-CK4) antibody to Cytokeratin 4 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Cytokeratin 4 + 5 + 6 + 8 + 10 + 13 + 18
Clonality: Monoclonal
Clone: ABT-CK4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR