Anti-Synaptophysin Mouse Monoclonal Antibody [clone: ABT-SYN]

Supplier: ANTIBODIES.COM LLC
A97802-100 A97802-50
77059-762EA 765.04 CAD
77059-762 77059-764
Anti-Synaptophysin Mouse Monoclonal Antibody [clone: ABT-SYN]
Antibodies
Mouse monoclonal (ABT-SYN) antibody to Synaptophysin for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Synaptophysin
Clonality: Monoclonal
Clone: ABT-SYN
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR