Anti-Claudin 3 Mouse Monoclonal Antibody [clone: ABT-CLD3]

Supplier: ANTIBODIES.COM LLC
A97074-100 A97074-50
77056-846EA 765.04 CAD
77056-846 77056-848
Anti-Claudin 3 Mouse Monoclonal Antibody [clone: ABT-CLD3]
Antibodies
Mouse monoclonal (ABT-CLD3) antibody to Claudin 3 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Claudin 3
Clonality: Monoclonal
Clone: ABT-CLD3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR