Anti-Claudin 4 Mouse Monoclonal Antibody [clone: ABT-CLD4]

Supplier: ANTIBODIES.COM LLC
A97073-100 A97073-50
77056-842EA 765.04 CAD
77056-842 77056-844
Anti-Claudin 4 Mouse Monoclonal Antibody [clone: ABT-CLD4]
Antibodies
Mouse monoclonal (ABT-CLD4) antibody to Claudin 4 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Claudin 4
Clonality: Monoclonal
Clone: ABT-CLD4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR