Anti-Claudin 7 Mouse Monoclonal Antibody [clone: ABT-CLD7]

Supplier: ANTIBODIES.COM LLC
A97072-100 A97072-50
77056-838EA 765.04 CAD
77056-838 77056-840
Anti-Claudin 7 Mouse Monoclonal Antibody [clone: ABT-CLD7]
Antibodies
Mouse monoclonal (ABT-CLD7) antibody to Claudin 7 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Claudin 7
Clonality: Monoclonal
Clone: ABT-CLD7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR