Anti-pan Cadherin Mouse Monoclonal Antibody [clone: ABT-CDHE]

Supplier: ANTIBODIES.COM LLC
A96686-100 A96686-50
77055-294EA 765.04 CAD
77055-294 77055-296
Anti-pan Cadherin Mouse Monoclonal Antibody [clone: ABT-CDHE]
Antibodies
Mouse monoclonal (ABT-CDHE) antibody to E Cadherin for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: pan Cadherin
Clonality: Monoclonal
Clone: ABT-CDHE
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR