Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: ABT-CK10]

Supplier: ANTIBODIES.COM LLC
A96673-100 A96673-50
77055-242EA 765.04 CAD
77055-242 77055-244
Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: ABT-CK10]
Antibodies
Mouse monoclonal (ABT-CK10) antibody to Cytokeratin 10 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Cytokeratin 10 + 13
Clonality: Monoclonal
Clone: ABT-CK10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR