Anti-Cytokeratin 6a Mouse Monoclonal Antibody [clone: ABT-CK6]

Supplier: ANTIBODIES.COM LLC
A96297-100 A96297-50
77053-738EA 765.04 CAD
77053-738 77053-740
Anti-Cytokeratin 6a Mouse Monoclonal Antibody [clone: ABT-CK6]
Antibodies
Mouse monoclonal (ABT-CK6) antibody to Cytokeratin 6 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Cytokeratin 6a
Clonality: Monoclonal
Clone: ABT-CK6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR