Anti-Cytokeratin 7+17 Mouse Monoclonal Antibody [clone: ABT-CK7]

Supplier: ANTIBODIES.COM LLC
A96296-50 A96296-100
77053-736EA 563.74 CAD
77053-736 77053-734
Anti-Cytokeratin 7+17 Mouse Monoclonal Antibody [clone: ABT-CK7]
Antibodies
Mouse monoclonal (ABT-CK7) antibody to Cytokeratin 7 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Cytokeratin 7+17
Clonality: Monoclonal
Clone: ABT-CK7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR