Anti-Cytokeratin 8+18 Mouse Monoclonal Antibody [clone: ABT-CK8]

Supplier: ANTIBODIES.COM LLC
A96295-100 A96295-50
77053-730EA 765.04 CAD
77053-730 77053-732
Anti-Cytokeratin 8+18 Mouse Monoclonal Antibody [clone: ABT-CK8]
Antibodies
Mouse monoclonal (ABT-CK8) antibody to Cytokeratin 8 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Cytokeratin 8+18
Clonality: Monoclonal
Clone: ABT-CK8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR