Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK13]

Supplier: ANTIBODIES.COM LLC
A95697-50 A95697-100
77051-338EA 563.74 CAD
77051-338 77051-336
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK13]
Antibodies
Mouse monoclonal (ABT-CK13) antibody to Cytokeratin 13 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-CK13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR