Anti-Myelin Basic Protein Mouse Monoclonal Antibody [clone: ABT-MBP]

Supplier: ANTIBODIES.COM LLC
A95520-100 A95520-50
77050-628EA 765.04 CAD
77050-628 77050-630
Anti-Myelin Basic Protein Mouse Monoclonal Antibody [clone: ABT-MBP]
Antibodies
Mouse monoclonal (ABT-MBP) antibody to Myelin Basic Protein for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: Myelin Basic Protein
Clonality: Monoclonal
Clone: ABT-MBP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR