Anti-Collagen III Mouse Monoclonal Antibody [clone: ABT-CIII]

Supplier: ANTIBODIES.COM LLC
A95291-100 A95291-50
77049-712EA 765.04 CAD
77049-712 77049-714
Anti-Collagen III Mouse Monoclonal Antibody [clone: ABT-CIII]
Antibodies
Mouse monoclonal (ABT-CIII) antibody to Collagen Type III for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: Collagen III
Clonality: Monoclonal
Clone: ABT-CIII
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR