Anti-Topoisomerase II alpha+Topoisomerase II beta Mouse Monoclonal Antibody [clone: ABT-TOPO]

Supplier: ANTIBODIES.COM LLC
A94715-50 A94715-100
77047-406EA 563.74 CAD
77047-406 77047-404
Anti-Topoisomerase II alpha+Topoisomerase II beta Mouse Monoclonal Antibody [clone: ABT-TOPO]
Antibodies
Mouse monoclonal (ABT-TOPO) antibody to Topoisomerase II alpha for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB
  • Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:1000

Type: Primary
Antigen: Topoisomerase II alpha+Topoisomerase II beta
Clonality: Monoclonal
Clone: ABT-TOPO
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR