Anti-Prostatic Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-PSAP]

Supplier: ANTIBODIES.COM LLC
A94698-100 A94698-50
77047-336EA 765.04 CAD
77047-336 77047-338
Anti-Prostatic Acid Phosphatase Mouse Monoclonal Antibody [clone: ABT-PSAP]
Antibodies
Mouse monoclonal (ABT-PSAP) antibody to Prostatic Acid Phosphatase for IHC with samples derived from Human.

  • Validated Applications: IHC
  • Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:20000

Type: Primary
Antigen: Prostatic Acid Phosphatase
Clonality: Monoclonal
Clone: ABT-PSAP
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR