Chromatography Columns, with Spherical Joint, PTFE Stopcock and Reservoir
Supplier: Chemglass
CG-1202-21 CG-1202-22 CG-1202-28 CG-1202-29 CG-1202-30 CG-1202-33 CG-1202-34 CG-1202-35 CG-1202-20 CG-1202-23 CG-1202-24 CG-1202-25 CG-1202-26 CG-1202-27
80067-120EA 443.11 CAD
80067-120 80067-122 80067-134 80067-136 80067-138 80067-144 80067-146 80067-148 80067-118 80067-124 80067-126 80067-128 80067-130 80067-132
Chromatography Columns, with Spherical Joint, PTFE Stopcock and Reservoir
Chromatography Columns
General purpose column has a PTFE stopcock, reservoir of the listed capacity and a top spherical socket joint.
  • Column is constructed using medium wall tubing
  • Glass wool (not supplied) is used to support the column packing
  • Lower end of the column has a round bottom
Can be used as a gravity eluted column or a flash column, when used with optional CG-1201 flow control adapter.