Cubelets® Clever Constructors Pack
Supplier: MODULAR ROBOTICS
Cubelets®
855165004864
470352-678PK 4581.99 CAD
470352-678
Cubelets® Clever Constructors Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The most popular Cubelets® educator pack.
  • Designed to be intuitive and screen-free for the youngest learners
  • Incorporate coding and computer science principles for older students
  • Additional Cubelets® support more advanced robot designs
  • Kit supports six student groups
  • Includes 78 Cubelets® (11 of 16 Cubelet types), and 12 brick adapters
  • Neatly stores away in one durable plastic tub
Perfect for classrooms and small makerspaces. Clever constructors is the most popular size and configuration for classroom educators. Build steerable robots and more with this perfect screen-free robotics tool. Foster vital thinking skills in your classroom, makerspace, library, or museum. Fascinating and fun, Cubelets® nurture your student's imagination, creativity, and inventive spirit. Cubelets® are the building blocks of better computational thinkers.
This quintessential Cubelets® collection has been expanded for six student groups. The Clever constructors pack facilitates easy adoption and high-ceiling academic activities. Includes 78 SENSE, THINK and ACT Cubelets®, brick adapters, 5-port charger, micro-USB charging cables and a new easy-to-organize storage tub. Every Cubelets® set and pack includes access to free standards-aligned lesson plans, printable activity cards, online professional development, and more.
Cubelets® magnetic faces make them simple to snap together and pull apart. Each Cubelet has a special function. What you connect, and how you connect them changes the behavior of your robot constructions: every combination is a new creation. Each Cubelet belongs to one of three categories: SENSE Cubelets® take in information from the robot's environment. THINK Cubelets® compute and change that information, and ACT Cubelets® turn that information into physical action. Connect at least one SENSE, THINK, and ACT Cubelet to create a robot construction. Inside of each block is a part of a program – as you build your robot you build the code that controls its behavior. It's hands-on coding.
Ordering information: Recommended accessories: (6) bluetooth hats.
Delivery information: Kit includes 78 Cubelets®:
6 battery, 6 passive, 6 inverse, 6 threshold, 12 distance, 6 brightness, 6 knob, 12 drive, 6 flashlight, 6 rotate and 6 bar graph. Plus: 12 brick adapters, 1×5-port charger, 5 charging cables and 1 storage tub.