Cubelets® Motivated Makers Pack
Supplier: MODULAR ROBOTICS
Cubelets®
855165004857
470352-684PK 3207.39 CAD
470352-684
Cubelets® Motivated Makers Pack
Educational Materials Physics Educational Materials Applied Physics Learning Activities
The perfect way to introduce STEM, robotics and computational thinking.
  • Designed to be intuitive and screen-free for the youngest learners
  • Incorporate coding and computer science principles for older students
  • Kit supports six student groups
  • Includes 52 Cubelets® (8 of 16 Cubelet types), and 24 brick adapters
  • Neatly stores away in one durable plastic tub
Designed for the needs of the youngest builders, the motivated makers is the perfect way to introduce your students to robotics, STEM, and computational thinking. This pack supports six working groups with screen-free explorations. Foster vital thinking skills in your classroom, makerspace, library, or museum. Fascinating and fun, Cubelets® nurture your student's imagination, creativity, and inventive spirit. Cubelets® are the building blocks of better computational thinkers.
Supporting six student groups, this pack includes 52 SENSE, THINK, and ACT Cubelets®. Also included are brick adapters, a 5-port charger, micro-USB charging cables, and a new, easy-to-organize storage tub. Every Cubelets® set and pack includes access to free standards-aligned lesson plans, printable activity cards, online professional development, and more.
Cubelets® magnetic faces make them simple to snap together and pull apart. Each Cubelet has a special function. What you connect, and how you connect them changes the behavior of your robot constructions: every combination is a new creation. Each Cubelet belongs to one of three categories: SENSE Cubelets® take in information from the robot's environment. THINK Cubelets® compute and change that information, and ACT Cubelets® turn that information into physical action. Connect at least one SENSE, THINK, and ACT Cubelet to create a robot construction. Inside of each block is a part of a program – as you build your robot you build the code that controls its behavior. It's hands-on coding.
Ordering information: Recommended accessories: (6) bluetooth hats.
Delivery information: Kit includes 52 Cubelets®:
6 battery, 6 passive, 6 inverse, 6 distance, 6 brightness, 6 drive, 6 flashlight, 6 rotate, and 4 knob. Plus: 12 stud brick adapters, 12 socket brick adapters, one 5-port charger, 5 charging cables and 1 storage tub.