Chromatography Columns, with Rodaviss® Joint and PTFE Stopcock
Supplier: Chemglass
CG-1192-01 CG-1192-04 CG-1192-05 CG-1192-08 CG-1192-13 CG-1192-20 CG-1192-21 CG-1192-24 CG-1192-25 CG-1192-26 CG-1192-29 CG-1192-30 CG-1192-31 CG-1192-03 CG-1192-07 CG-1192-10 CG-1192-11 CG-1192-12 CG-1192-15 CG-1192-16 CG-1192-19
80066-752EA 396.57 CAD
80066-752 80066-758 80066-760 80066-766 80066-776 80066-790 80066-792 80066-798 80066-800 80066-802 80066-808 80066-810 80066-812 80066-756 80066-764 80066-770 80066-772 80066-774 80066-780 80066-782 80066-788
Chromatography Columns, with Rodaviss® Joint and PTFE Stopcock
Chromatography Columns
General purpose column has a PTFE stopcock and a top Rodaviss® outer joint.
  • Column is constructed using medium wall tubing
  • Glass wool (not supplied) is used to support column packing
  • Lower end of the column has a round bottom
Can be used as a gravity eluted column or a flash column, when used with optional CG-1195 flow controller and CG-1194 reservoir.
Connections between the column, reservoir and flow control adapter are made using Rodaviss® joints. The unique design of the Rodaviss® joint affords a grease free seal while holding glassware together without clamps. Column is supplied complete with a CG-182 open top cap and O-ring.