Supplier: Chemglass
CG-1187-01 CG-1187-02 CG-1187-04 CG-1187-07 CG-1187-08 CG-1187-10 CG-1187-19 CG-1187-20 CG-1187-21 CG-1187-29 CG-1187-31 CG-1187-03 CG-1187-11 CG-1187-16 CG-1187-06 CG-1187-12 CG-1187-15
80066-544EA 340.52 CAD
80066-544 80066-546 80066-550 80066-556 80066-558 80066-562 80066-580 80066-582 80066-584 80066-600 80066-604 80066-548 80066-564 10170-364 10170-806 10170-808 10170-810
Chromatography Columns
Gravity eluted column has a PTFE stopcock and reinforced beaded top, with a sealed-in coarse porosity fritted disc to support column packing.
  • Column is constructed using medium wall tubing
  • Lower end of the column has a round bottom