Chromatography Columns, with PTFE Stopcock
Supplier: Chemglass
CG-1186-04 CG-1186-06 CG-1186-08 CG-1186-12 CG-1186-15 CG-1186-19 CG-1186-20 CG-1186-21 CG-1186-29 CG-1186-31 CG-1186-01 CG-1186-02 CG-1186-03 CG-1186-07 CG-1186-10 CG-1186-11 CG-1186-16
80066-488EA 437.54 CAD
80066-488 80066-492 80066-496 80066-504 80066-510 80066-518 80066-520 80066-522 80066-538 80066-542 80066-482 80066-484 80066-486 80066-494 80066-500 80066-502 80066-512
Chromatography Columns, with PTFE Stopcock
Chromatography Columns
General purpose gravity eluted column has a PTFE stopcock and reinforced beaded top.
  • Column is constructed using medium wall tubing
  • Glass wool (not supplied) is used to support column packing
  • Lower end of the column has a round bottom