Mouse Pancreatic Lipase/PTL ELISA Kit
Supplier: ANTIBODIES.COM LLC
Mouse Pancreatic Lipase/PTL ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Pancreatic Lipase/PTL in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 3 to 48 ng/ml
- Sensitivity: 0.14 ng/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Pancreatic Lipase/PTL ELISA kit (A313719) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Pancreatic Lipase/PTL in serum, plasma or other biological fluids. An antibody specific for Pancreatic Lipase/PTL has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Pancreatic Lipase/PTL present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Pancreatic Lipase/PTL antibody, which also binds the Pancreatic Lipase/PTL present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Pancreatic Lipase/PTL antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Pancreatic Lipase/PTL antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Pancreatic Lipase/PTL captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Pancreatic Lipase/PTL in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...