Mouse NMU ELISA Kit

Supplier: Antibodies.com LLC
A313726-96
77174-378EA 854.39 CAD
77174-378
Mouse NMU ELISA Kit
Assays ELISAs
Mouse NMU ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse NMU in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 4 to 64 ng/ml
  • Sensitivity: 0.224 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse nmU ELISA kit (A313726) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse nmU in serum, plasma or other biological fluids. An antibody specific for nmU has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the nmU present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti- nmU antibody, which also binds the nmU present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti- nmU antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti- nmU antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of nmU captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of nmU in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR