Mouse LGALS3BP ELISA Kit

Supplier: Antibodies.com LLC
A313351-96
77173-632EA 854.39 CAD
77173-632
Mouse LGALS3BP ELISA Kit
Assays ELISAs
Mouse LGALS3BP ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse LGALS3BP in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 1.5 to 24 ng/ml
  • Sensitivity: 0.052 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse LGALS3BP ELISA kit (A313351) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse LGALS3BP in serum, plasma or other biological fluids. An antibody specific for LGALS3BP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the LGALS3BP present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-LGALS3BP antibody, which also binds the LGALS3BP present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-LGALS3BP antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-LGALS3BP antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of LGALS3BP captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of LGALS3BP in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR