Mouse IGFBP7 ELISA Kit

Supplier: Antibodies.com LLC
A313231-96
77173-392EA 854.39 CAD
77173-392
Mouse IGFBP7 ELISA Kit
Assays ELISAs
Mouse IGFBP7 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IGFBP7 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 50 to 800 ng/L
  • Sensitivity: 3.32 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse IGFBP7 ELISA kit (A313231) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse IGFBP7 in serum, plasma or other biological fluids. An antibody specific for IGFBP7 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IGFBP7 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-IGFBP7 antibody, which also binds the IGFBP7 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IGFBP7 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IGFBP7 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of IGFBP7 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of IGFBP7 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR